Skip to Content
Merck
All Photos(1)

Key Documents

8.08255

Sigma-Aldrich

Trichloromethylsilane

for synthesis

Synonym(s):

Trichloromethylsilane, Methyltrichlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$55.40
1 L
$132.00

$55.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$55.40
1 L
$132.00

About This Item

Empirical Formula (Hill Notation):
CH3Cl3Si
CAS Number:
Molecular Weight:
149.48
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-902-6
NACRES:
NA.22

$55.40


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

189 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

>500 °C

expl. lim.

5.5-10.4 % (v/v)

mp

-90 °C

transition temp

flash point -1 °C

density

1.27 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)C

InChI

1S/CH3Cl3Si/c1-5(2,3)4/h1H3

InChI key

JLUFWMXJHAVVNN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.271 - 1.275
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service