This product is 1,2,3,4-Tetrahydro-1-naphthalenone (α-Tetralone) is an enantiomer of 1,2,3,4-Tetrahydro-2-naphthoic acid (β-Tetralone). The requested information is not currently available.
It may be helpful to check the available literature on this topic.
8.08203
1,2,3,4-Tetrahydro-1-naphthalenone
for synthesis
Synonym(s):
1,2,3,4-Tetrahydro-1-naphthalenone, α-Tetralone
Select a Size
About This Item
Recommended Products
Quality Level
Assay
≥96.0% (GC)
form
liquid
potency
810 mg/kg LD50, oral (Rat)
>2200 mg/kg LD50, skin (Rabbit)
bp
113-116 °C/8 hPa
mp
5-6 °C
transition temp
flash point >110 °C
density
1.10 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O=C1CCCc2c1cccc2
InChI
1S/C10H10O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6H,3,5,7H2
InChI key
XHLHPRDBBAGVEG-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.092 - 1.095
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How to convert to 1234 tetra hydro 2 napthalenone
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service