Skip to Content
Merck
All Photos(1)

Key Documents

8.07486

Sigma-Aldrich

Polyethylene glycol

600, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 600, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 600, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$66.30
5 L
$257.00

$66.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
$66.30
5 L
$257.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
13111021
EC Index Number:
500-038-2
NACRES:
NA.22

$66.30


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

>320 °C (DIN 51794)

potency

28000 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

17-22 °C

transition temp

flash point 260 °C

density

1.13 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polyethylene glycol 600 for synthesis. CAS 25322-68-3, pH 5 - 7 (100 g/l, H₂O, 20 °C).

Analysis Note

Density (d 20 °C/ 4 °C): 1.126 - 1.128
Hydroxyl value: 178 - 197
Average molecular mass: 570 - 630
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the kinetic viscosity in cSt at 37°C?

    1 answer
    1. The kinetic viscosity is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service