Skip to Content
Merck
All Photos(1)

Key Documents

8.07270

Sigma-Aldrich

Phosphorus (red)

(stabilised) for synthesis

Synonym(s):

Phosphorus (red)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$62.20
500 G
$114.00
2.5 KG
$335.00

$62.20


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$62.20
500 G
$114.00
2.5 KG
$335.00

About This Item

Empirical Formula (Hill Notation):
P
CAS Number:
Molecular Weight:
30.97
MDL number:
UNSPSC Code:
12141909
EC Index Number:
231-768-7
NACRES:
NA.22

$62.20


Estimated to ship on31 May 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

form

powder

autoignition temp.

260 °C

potency

>15000 mg/kg LD50, oral (Rat)

mp

585 °C

density

2.2 g/cm3

bulk density

1100 kg/m3

storage temp.

2-30°C

SMILES string

[P]

InChI

1S/P

InChI key

OAICVXFJPJFONN-UHFFFAOYSA-N

Application


  • Design of red phosphorus nanostructured electrode for fast-charging lithium-ion batteries: This study proposes red phosphorus as an ideal anode material for fast-charging lithium-ion batteries, demonstrating a red phosphorus-carbon nanocomposite with excellent fast-charging capability and high energy density. (Sun et al., 2019).

  • Elemental red phosphorus-based photocatalysts for environmental remediation: A review of low-cost and environmentally benign elemental red phosphorus as a new class of photocatalysts with tunable bandgaps for visible-light response, used in environmental remediation. (Wu et al., 2021).

  • Atomically Dispersed Au-Assisted C–C Coupling on Red Phosphorus for CO2 Photoreduction to C2H6: This study explores the use of atomically dispersed gold on red phosphorus for the photocatalytic reduction of CO2 to ethane, highlighting the potential of red phosphorus in photocatalytic applications. (Ou et al., 2022).

  • Rapid biofilm eradication on bone implants using red phosphorus and near‐infrared light: This research reports on the use of red phosphorus for rapid biofilm eradication on bone implants when exposed to near-infrared light, presenting a new method for medical applications. (Tan et al., 2018).

Analysis Note

Assay P: ≥ 97.0 %

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the size of the phosphorus powder? Can we get about 10 microns?

    1 answer
    1. The size of this powder is not determined. Unfortunately, there is no red phosphorus powder product currently offered that is 10 microns.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service