Skip to Content
Merck
All Photos(1)

Key Documents

8.07020

Sigma-Aldrich

Aminopropanol

(mixture of isomers) for synthesis

Synonym(s):

Aminopropanol, Aminoisopropyl alcohol, Hydroxypropylamine, Isopropanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$48.90
1 L
$67.20
190 KG
$3,950.00

$48.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$48.90
1 L
$67.20
190 KG
$3,950.00

About This Item

Linear Formula:
H2NCH2CH(OH)CH3
CAS Number:
Molecular Weight:
75.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-162-7
NACRES:
NA.22

$48.90


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

1.93 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

335 °C

potency

2700 mg/kg LD50, oral (Rat)

expl. lim.

1.9-10.4 % (v/v)

pH

11.3 (20 °C, 10 g/L in H2O)

bp

160 °C

mp

2 °C

transition temp

flash point 71 °C

density

0.962 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCC(O)C

InChI

1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3

InChI key

HXKKHQJGJAFBHI-UHFFFAOYSA-N

Analysis Note

Assay isomers (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.959 - 0.961
1-amino-2-propanol (GC, area%): ≥ 90.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service