Skip to Content
Merck
All Photos(1)

Key Documents

8.07006

Sigma-Aldrich

2-Phenylethanol

for synthesis

Synonym(s):

2-Phenylethanol, Phenethyl alcohol, Benzylcarbinol

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$35.80
250 ML
$50.50
1 L
$136.00
25 KG
$1,230.00

$35.80


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$35.80
250 ML
$50.50
1 L
$136.00
25 KG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-456-2
NACRES:
NA.22

$35.80


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

0.08 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

410 °C

potency

2230 mg/kg LD50, oral (Rat)
300-2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-11.9 % (v/v)

pH

6-7 (20 °C, 20 g/L in H2O)

bp

219 °C/1013 hPa

mp

-27 °C

transition temp

flash point 101.5 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCc1ccccc1

InChI

1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

WRMNZCZEMHIOCP-UHFFFAOYSA-N

Application

  • The bacteriostatic activity of 2-phenylethanol derivatives correlates with membrane binding affinity: This study examines how the bacteriostatic properties of 2-phenylethanol derivatives are influenced by their affinity for binding to membranes, providing insights into their antimicrobial effectiveness (IS Kleinwächter et al., 2021).
  • Anti-depressive-like effect of 2-phenylethanol inhalation in mice: This study explores the neuropsychological effects of inhaling 2-phenylethanol, suggesting potential therapeutic applications for depression (H Ueno et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.019 - 1.020
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service