Skip to Content
Merck
All Photos(1)

Key Documents

8.06210

Sigma-Aldrich

1-Bromonaphthalene

for synthesis

Synonym(s):

1-Bromonaphthalene, α-Naphthyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$69.30
500 ML
$295.00
2.5 L
$1,300.00

$69.30


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$69.30
500 ML
$295.00
2.5 L
$1,300.00

About This Item

Empirical Formula (Hill Notation):
C10H7Br
CAS Number:
Molecular Weight:
207.07
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-965-2
NACRES:
NA.22

$69.30


Estimated to ship on31 May 2025


Request a Bulk Order

vapor pressure

0.013 hPa ( 20 °C)

Quality Level

form

liquid

potency

810 mg/kg LD50, oral (Rat)

bp

281 °C/1013 hPa

transition temp

flash point 67 °C

density

1.48 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Brc1c2c(ccc1)cccc2

InChI

1S/C10H7Br/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

DLKQHBOKULLWDQ-UHFFFAOYSA-N

Analysis Note

Assay
- GC, area%:
≥ 95 %
- 2-Bromonaphthaline: ≤ 5 %
Density (d 20 °C/ 4 °C): 1.478 - 1.487
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service