Skip to Content
Merck
All Photos(1)

Key Documents

8.05891

Sigma-Aldrich

3-Methoxypropionitrile

for synthesis

Synonym(s):

3-Methoxypropionitrile, 2-Methoxyethyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$58.90
500 ML
$201.00

$58.90


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$58.90
500 ML
$201.00

About This Item

Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
MDL number:
UNSPSC Code:
12352117
EC Index Number:
203-790-7
NACRES:
NA.22

$58.90


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

2.3 hPa ( 30 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

390 °C (DIN 51794)

potency

4130 mg/kg LD50, oral (Rat)
>9400 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-15.3 % (v/v)

pH

6.8 (25 °C, 111 g/L in H2O)

bp

164-165 °C/1013 hPa

mp

-63 °C

transition temp

flash point 66 °C

solubility

540 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#CCCOC

InChI

1S/C4H7NO/c1-6-4-2-3-5/h2,4H2,1H3

InChI key

OOWFYDWAMOKVSF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.938 - 0.940
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service