Skip to Content
Merck
All Photos(1)

Key Documents

8.05792

Sigma-Aldrich

Methacrylamide

for synthesis

Synonym(s):

Methacrylamide, 2-Methyl-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$95.80

$95.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
$95.80

About This Item

Linear Formula:
CH2=C(CH3)CONH2
CAS Number:
Molecular Weight:
85.10
MDL number:
UNSPSC Code:
12352111
EC Index Number:
201-202-3
NACRES:
NA.22

$95.80


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

0.0015 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

autoignition temp.

510 °C

potency

1815 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

expl. lim.

2-15 % (v/v)

pH

7.1 (20 °C, 100 g/L in H2O)

bp

215 °C/1013 hPa

mp

106-112 °C

solubility

202 g/L

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

NC(=O)C(=C)C

InChI

1S/C4H7NO/c1-3(2)4(5)6/h1H2,2H3,(H2,5,6)

InChI key

FQPSGWSUVKBHSU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 109 °C
Melting range (upper value): ≤ 111 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service