Skip to Content
Merck
All Photos(1)

Key Documents

8.05660

Sigma-Aldrich

Lithium

(sticks) (protective liquid: paraffin oil) for synthesis

Synonym(s):

Lithium

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$441.00

$441.00


Available to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$441.00

About This Item

Empirical Formula (Hill Notation):
Li
CAS Number:
Molecular Weight:
6.94
MDL number:
UNSPSC Code:
12141803
EC Index Number:
231-102-5
NACRES:
NA.22

$441.00


Available to ship on28 March 2025Details


Request a Bulk Order

vapor pressure

1 hPa ( 723 °C)

Quality Level

form

solid

mp

179 °C

density

0.534 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Li]

InChI

1S/Li

InChI key

WHXSMMKQMYFTQS-UHFFFAOYSA-N

Analysis Note

Assay of Li: ≥ 99.0 %

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service