Skip to Content
Merck
All Photos(2)

Key Documents

8.03912

Sigma-Aldrich

Iron(III) acetylacetonate

for synthesis

Synonym(s):

Iron(III) acetylacetonate, Tris(acetylacetonato)iron(III), Tris(2,4-pentanedionato)iron(III)

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$158.00

$158.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
$158.00

About This Item

Empirical Formula (Hill Notation):
C15H21FeO6
CAS Number:
Molecular Weight:
353.17
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-853-5
NACRES:
NA.22

$158.00


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

2.6 hPa ( 110 °C)

Quality Level

form

powder

potency

1872 mg/kg LD50, oral (Rat)

mp

180-183 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[Fe+3].[O-]\C(=C/C(=O)C)\C.[O-]\C(=C/C(=O)C)\C.[O-]\C(=C/C(=O)C)\C

InChI

1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

AQBLLJNPHDIAPN-LNTINUHCSA-K

Application

  • UV-Vis Spectroscopy and Electrochemical Analysis: Iron(III) acetylacetonate is used in UV-Vis spectroscopy and electrochemical studies to explore its role in dye-sensitized solar cells (DSSCs), focusing on the impact of aromatic thienyl groups on the electronic properties of tris(β-diketonato)iron(III) complexes (Conradie MM, 2022).
  • One-Step Synthesis of Nanomaterials: The compound facilitates a one-step synthesis route to produce Fe2O3 and FeSe2 nanoparticles on carbon sheets, enhancing lithium storage capabilities in batteries (Wei D et al., 2022).

Analysis Note

Assay (ex Fe): ≥ 99,0 %(m)
Identity (IR): conforms

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service