Skip to Content
Merck
All Photos(1)

Key Documents

8.03580

Sigma-Aldrich

1,4-Dimethylpiperazine

for synthesis

Synonym(s):

1,4-Dimethylpiperazine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$61.40

$61.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$61.40

About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-412-0
NACRES:
NA.22

$61.40


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

20 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

180 °C

potency

2800 mg/kg LD50, oral (Rat)
3000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.8-9.6 % (v/v)

pH

11.4 ( in H2O)

mp

-1 °C

transition temp

flash point 22 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(CCN(CC1)C)C

InChI

1S/C6H14N2/c1-7-3-5-8(2)6-4-7/h3-6H2,1-2H3

InChI key

RXYPXQSKLGGKOL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.849 - 0.851
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service