Skip to Content
Merck
All Photos(2)

Key Documents

8.03573

Sigma-Aldrich

2,6-Diamino-4-phenyl-1,3,5-triazine

for synthesis

Synonym(s):

2,6-Diamino-4-phenyl-1,3,5-triazine, Benzoguanamine, 2,4-Diamino-6-phenyl-s-triazine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$54.00
1 KG
$156.00

$54.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 G
$54.00
1 KG
$156.00

About This Item

Empirical Formula (Hill Notation):
C9H9N5
CAS Number:
Molecular Weight:
187.20
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-095-6
NACRES:
NA.22

$54.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

form

powder

potency

1470 mg/kg LD50, oral (Rat)

pH

6.5 (20 °C, 0.3 g/L in H2O)

mp

224-227 °C

transition temp

flash point >270 °C

solubility

0.3 g/L

density

1.425 g/cm3 at 15 °C

bulk density

680 kg/m3

storage temp.

2-30°C

SMILES string

Nc1nc(nc(n1)c2ccccc2)N

InChI

1S/C9H9N5/c10-8-12-7(13-9(11)14-8)6-4-2-1-3-5-6/h1-5H,(H4,10,11,12,13,14)

InChI key

GZVHEAJQGPRDLQ-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 224 °C
Melting range (upper value): ≤ 228 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service