Skip to Content
Merck
All Photos(1)

Key Documents

8.03405

Sigma-Aldrich

n-Decane

for synthesis

Synonym(s):

n-Decane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$55.20
250 ML
$111.00
1 L
$431.00

$55.20


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$55.20
250 ML
$111.00
1 L
$431.00

About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
MDL number:
UNSPSC Code:
12352001
EC Index Number:
204-686-4
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
174 °C/1013 mbar
vapor pressure:
1 hPa ( 16 °C)

$55.20


Estimated to ship on02 June 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

1 hPa ( 16 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

208 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

0.7-5.4 % (v/v)

dilution

(for synthesis)

kinematic viscosity

<7 cSt(40 °C)

bp

174 °C/1013 mbar

mp

-30 °C

transition temp

flash point 51 °C

density

0.73 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCC)CCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Application

  • Molecular Dynamics in Surfactant Research: Dong et al. conducted molecular dynamics simulations to study the effects of surfactants on methane and n-decane interactions. This research provides insights into the applications of n-decane in enhancing the performance of surfactants in oil recovery and environmental remediation processes (Dong et al., 2024).
  • Interfacial Adsorption Studies: Ruwoldt et al. explored the interfacial adsorption properties of oil-soluble kraft lignin and its ability to stabilize water-in-oil emulsions, where n-decane plays a crucial role as a model hydrocarbon solvent, contributing to understanding emulsion stability and lignin-solvent interactions in industrial applications (Ruwoldt et al., 2024).
  • Semiconductor Strain Gauges in Geotechnical Engineering: Mikhaltsevitch and Lebedev utilized semiconductor strain gauges to measure the effective stress coefficient for the elastic moduli of sandstone. N-decane could potentially be used as a fluid medium in these experiments to simulate natural hydrocarbon environments, enhancing the accuracy of geophysical assessments (Mikhaltsevitch et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.729 - 0.730
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service