Skip to Content
Merck
All Photos(1)

Key Documents

8.03284

Sigma-Aldrich

Dimethyl sulfone

for synthesis

Synonym(s):

Dimethyl sulfone, Methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO2
CAS Number:
Molecular Weight:
94.13
MDL number:
UNSPSC Code:
12352119
EC Index Number:
200-665-9
Pricing and availability is not currently available.

Assay

≥99.0% (GC)

form

crystals

potency

17000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

238 °C/1013 hPa

mp

107-110 °C

transition temp

flash point 143 °C

bulk density

880 kg/m3

storage temp.

2-30°C

InChI

1S/C2H6O2S/c1-5(2,3)4/h1-2H3

InChI key

HHVIBTZHLRERCL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 107 °C
Melting range (upper value): ≤ 110 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service