Skip to Content
Merck
All Photos(2)

Key Documents

8.03257

Sigma-Aldrich

N-Phenyl-2,2′-iminodiethanol

for synthesis

Synonym(s):

N-Phenyl-2,2′-iminodiethanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$60.90
1 KG
$269.00

$60.90


Estimated to ship on20 May 2025



Select a Size

Change View
100 G
$60.90
1 KG
$269.00

About This Item

Linear Formula:
C6H5N(CH2CH2OH)2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-368-5
NACRES:
NA.22

$60.90


Estimated to ship on20 May 2025


vapor pressure

<1 hPa ( 20 °C)

Quality Level

autoignition temp.

380 °C (DIN 51794)

potency

3400 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

8.2 (50 °C, 3 g/L in H2O)

mp

54-59 °C

transition temp

flash point 200 °C

solubility

3 g/L

density

1.1 g/cm3 at 60 °C

storage temp.

2-30°C

SMILES string

N(CCO)(CCO)c1ccccc1

InChI

1S/C10H15NO2/c12-8-6-11(7-9-13)10-4-2-1-3-5-10/h1-5,12-13H,6-9H2

InChI key

OJPDDQSCZGTACX-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 58 °C
Identity (IR): passes test

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service