Skip to Content
Merck
All Photos(1)

Key Documents

8.02641

Sigma-Aldrich

5-Chloro-2-methylaniline

for synthesis

Synonym(s):

5-Chloro-2-methylaniline, 4-Chloro-2-toluidine, 5-Chloro-o-toluidine, 2-Amino-4-chlorotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$355.00

$355.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$355.00

About This Item

Linear Formula:
5-(Cl)-2-(CH3)C6H3NH2
CAS Number:
Molecular Weight:
141.60
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-452-6
NACRES:
NA.22

$355.00


Estimated to ship on02 June 2025


Request a Bulk Order

vapor pressure

0.45 hPa ( 50 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

>540 °C

potency

464 mg/kg LD50, oral (Rat)

bp

237 °C/1013 hPa

mp

22 °C

transition temp

flash point 160 °C

density

1.17 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1cc(c(cc1)C)N

InChI

1S/C7H8ClN/c1-5-2-3-6(8)4-7(5)9/h2-4H,9H2,1H3

InChI key

WRZOMWDJOLIVQP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 24 °C
Melting range (upper value): ≤ 28 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service