Skip to Content
Merck
All Photos(2)

Key Documents

8.02528

Sigma-Aldrich

Cobalt(III) acetylacetonate

for synthesis

Synonym(s):

Cobalt(III) acetylacetonate, Tris(acetylacetonato)cobalt(III), Tris(2,4-pentanedionato)cobalt(III)

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$151.00

$151.00


Estimated to ship on16 April 2025



Select a Size

Change View
50 G
$151.00

About This Item

Empirical Formula (Hill Notation):
C15H21CoO6
CAS Number:
Molecular Weight:
356.26
MDL number:
UNSPSC Code:
12352103
EC Index Number:
244-527-6
NACRES:
NA.22

$151.00


Estimated to ship on16 April 2025


Quality Level

mp

196-202 °C (decomposition)

solubility

3 g/L

bulk density

830 kg/m3

storage temp.

2-30°C

SMILES string

[Co](O\C(=C/C(=O)C)\C)(O\C(=C/C(=O)C)\C)O\C(=C/C(=O)C)\C

InChI

1S/3C5H8O2.Co/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

RHCQEPWEBDOALW-LNTINUHCSA-K

Analysis Note

Assay at Co: ≥ 98.0 %
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service