Skip to Content
Merck
All Photos(1)

Key Documents

8.02180

Sigma-Aldrich

Ethyl decanoate

for synthesis

Synonym(s):

Ethyl decanoate, Ethyl caprate, Decanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$36.00
250 ML
$93.30

$36.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$36.00
250 ML
$93.30

About This Item

Linear Formula:
CH3(CH2)8COOC2H5
CAS Number:
Molecular Weight:
200.32
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-761-9
NACRES:
NA.22

$36.00


Estimated to ship on21 May 2025


Request a Bulk Order

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

310 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7 % (v/v)

bp

244-246 °C/1013 hPa

mp

-20 °C

transition temp

flash point 104 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CCCCCCCCC

InChI

1S/C12H24O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h3-11H2,1-2H3

InChI key

RGXWDWUGBIJHDO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.863 - 0.864
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service