Skip to Content
Merck
All Photos(1)

Key Documents

8.01917

Sigma-Aldrich

Pyrocatechol monoethyl ether

for synthesis

Synonym(s):

Pyrocatechol monoethyl ether, Guaethol, 2-Ethoxyphenol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$108.00

$108.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$108.00

About This Item

Linear Formula:
2-(C2H5O)C6H4OH
CAS Number:
Molecular Weight:
138.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-358-5
NACRES:
NA.22

$108.00


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

1 hPa ( 45 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

bp

214-216 °C/1013 hPa

mp

24-28 °C

transition temp

flash point 86 °C

solubility

9 g/L

density

1.10 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)c1c(cccc1)O

InChI

1S/C8H10O2/c1-2-10-8-6-4-3-5-7(8)9/h3-6,9H,2H2,1H3

InChI key

MOEFFSWKSMRFRQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service