Skip to Content
Merck
All Photos(1)

Key Documents

8.01553

Sigma-Aldrich

Butyl 2,3-epoxypropyl ether

for synthesis

Synonym(s):

Butyl 2,3-epoxypropyl ether, Glycidol butyl ether, Butyl glycidyl ether, BGE, 1,2-Epoxy-3-butoxypropane

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$59.10

$59.10


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$59.10

About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
MDL number:
UNSPSC Code:
12352112
EC Index Number:
219-376-4
NACRES:
NA.22

$59.10


Estimated to ship on18 May 2025


Request a Bulk Order

vapor pressure

5 hPa ( 20 °C)

Quality Level

Assay

≥94.0% (GC)

form

liquid

autoignition temp.

215 °C

potency

1660 mg/kg LD50, oral (Rat)
2520 mg/kg LD50, skin (Rabbit)

bp

163-165 °C/1013 mbar

transition temp

flash point 43 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1C(C1)COCCCC

InChI

1S/C7H14O2/c1-2-3-4-8-5-7-6-9-7/h7H,2-6H2,1H3

InChI key

YSUQLAYJZDEMOT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.911 - 0.915
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service