Skip to Content
Merck
All Photos(1)

Key Documents

8.01532

Sigma-Aldrich

1,4-Butanediol

for synthesis

Synonym(s):

1,4-Butanediol, 1,4-Butylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$73.90
2.5 L
$146.00
25 L
$645.00

$73.90


Estimated to ship on29 May 2025



Select a Size

Change View
1 L
$73.90
2.5 L
$146.00
25 L
$645.00

About This Item

Linear Formula:
HO(CH2)4OH
CAS Number:
Molecular Weight:
90.12
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-786-5
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
228 °C/1013 hPa
vapor pressure:
<0.1 hPa ( 20 °C)

$73.90


Estimated to ship on29 May 2025


grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

385 °C

potency

1525 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.95-18.3 % (v/v)

dilution

(for synthesis)

pH

7-8 (20 °C, 500 g/L in H2O)

kinematic viscosity

83.2 cSt(20 °C)

bp

228 °C/1013 hPa

mp

19.5-20.0 °C

transition temp

flash point 115 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCO

InChI

1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

WERYXYBDKMZEQL-UHFFFAOYSA-N

Application

  • Non-Isocyanate Route Synthesis of Polyester Polyureas: Investigation into the synthesis of polyester polyureas through a non-isocyanate route using 1,4-Butanediol, focusing on developing polymers with combined properties for industrial and environmental applications (Zheng L et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.015 - 1.016
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

273.2 °F - closed cup

Flash Point(C)

134 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service