Skip to Content
Merck
All Photos(1)

Key Documents

8.00889

Sigma-Aldrich

Methyl formate

for synthesis

Synonym(s):

Methyl formate, Formic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$37.50
1 L
$59.00
2.5 L
$115.00

$37.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$37.50
1 L
$59.00
2.5 L
$115.00

About This Item

Empirical Formula (Hill Notation):
C2H4O2
CAS Number:
Molecular Weight:
60.05
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-481-7
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
31.5 °C/1013 hPa
vapor pressure:
644 hPa ( 20 °C)

$37.50


Estimated to ship on30 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

644 hPa ( 20 °C)

Assay

95.0-97.0% (GC)

form

liquid

autoignition temp.

450 °C

potency

1500 mg/kg LD50, oral (Rat)
>4000 mg/kg LD50, skin (Rat)

expl. lim.

5-23 % (v/v)

dilution

(for synthesis)

pH

4-5 (20 °C, 200 g/L in H2O)

bp

31.5 °C/1013 hPa

mp

-100.4 °C

transition temp

flash point -32 °C

solubility

300 g/L

density

0.968 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C=O

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): 95.0 - 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.967 - 0.971
Methanol (GC, area%): 3.0 - 5.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service