Skip to Content
Merck
All Photos(1)

Key Documents

8.00786

Sigma-Aldrich

Butyl 3-pyridinecarboxylate

for synthesis

Synonym(s):

Butyl 3-pyridinecarboxylate, Butyl nicotinate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$132.00

$132.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$132.00

About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
12352108
EC Index Number:
230-064-7
NACRES:
NA.22

$132.00


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

oil

bp

122-123 °C/11 hPa

transition temp

flash point 109 °C

density

1.05 g/cm3

storage temp.

2-30°C

SMILES string

n1cc(ccc1)C(=O)OCCCC

InChI

1S/C10H13NO2/c1-2-3-7-13-10(12)9-5-4-6-11-8-9/h4-6,8H,2-3,7H2,1H3

InChI key

DQULIMIQTCDUAN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.049 - 1.053
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F

Flash Point(C)

109 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service