Skip to Content
Merck
All Photos(1)

Key Documents

8.00758

Sigma-Aldrich

2-Ethylhexanoic acid

for synthesis

Synonym(s):

2-Ethylhexanoic acid, 3-Heptanecarboxylic acid, α-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$39.40
1 L
$65.60

$39.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$39.40
1 L
$65.60

About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-743-6
NACRES:
NA.22

$39.40


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

0.04 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

310 °C

potency

3640 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-6.7 % (v/v)

pH

3 (20 °C, 1.4 g/L in H2O)

bp

226-229 °C/1013 hPa

mp

-59 °C

transition temp

flash point 114 °C

solubility

1.4 g/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)C(CCCC)CC

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.905 - 0.907
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service