Skip to Content
Merck
All Photos(1)

Key Documents

8.00753

Sigma-Aldrich

Sebacic acid

for synthesis

Synonym(s):

Sebacic acid, Decanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$41.00
500 G
$114.00
1 KG
$183.00
50 KG
$1,960.00

$41.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$41.00
500 G
$114.00
1 KG
$183.00
50 KG
$1,960.00

About This Item

Linear Formula:
HOOC(CH2)8COOH
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-845-5
NACRES:
NA.22

$41.00


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

1.3 hPa ( 183 °C)

Quality Level

Assay

≥98.0% (GC)

form

powder

potency

3400-14500 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

bp

295 °C/133 hPa

mp

130-133 °C

solubility

1 g/L

density

1.21 g/cm3

bulk density

600‑620 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCCCC(=O)O

InChI

1S/C10H18O4/c11-9(12)7-5-3-1-2-4-6-8-10(13)14/h1-8H2,(H,11,12)(H,13,14)

InChI key

CXMXRPHRNRROMY-UHFFFAOYSA-N

Related Categories

Application

  • Biomedical applications: Sebacic acid was utilized in the fabrication of electrospun composite PLLA-PPSB nanofiber nerve conduits, which are designed for the repair and regeneration of peripheral nerve defects. This application leverages sebacic acid′s biocompatibility and biodegradability in advanced healthcare materials (Dai et al., 2024).
  • Polymer gel technologies: Research demonstrates the ability to tailor the properties of poly(vinyl alcohol) gels through sebacic acid decoration, optimizing characteristics such as porosity and elasticity for various industrial applications, including medical and consumer products (Bandelli et al., 2024).
  • High-performance materials: The synthesis and characterization of epoxy vitrimers incorporating sebacic acid have been explored. These vitrimers exhibit enhanced glass transition temperatures, making them suitable for applications requiring high thermal stability and mechanical properties in sectors such as automotive and aerospace (Dai et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 131 °C
Melting range (upper value): ≤ 134 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service