Skip to Content
Merck
All Photos(1)

Key Documents

8.00649

Sigma-Aldrich

3-Nitrophthalic acid

for synthesis

Synonym(s):

3-Nitrophthalic acid, 3-Nitrobenzene-1,2-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(NO2)C6H3-1,2-(COOH)2
CAS Number:
Molecular Weight:
211.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-030-8
Pricing and availability is not currently available.

Quality Level

form

powder

mp

210-215 °C

solubility

20 g/L

storage temp.

2-30°C

InChI

1S/C8H5NO6/c10-7(11)4-2-1-3-5(9(14)15)6(4)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

KFIRODWJCYBBHY-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service