Skip to Content
Merck
All Photos(2)

Key Documents

8.00589

Sigma-Aldrich

Methacrylic acid alkylester (mixture of C₁₂-C₁₆)

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

Methacrylic acid alkylester (mixture of C₁₂-C₁₆), Dodecyl 2-methylpropenoate, Lauryl methacrylate, Methacrylic acid dodecyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$56.70
500 ML
$82.70

$56.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 ML
$56.70
500 ML
$82.70

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352108
EC Index Number:
292-094-7
NACRES:
NA.22

$56.70


0 left in stock

Request a Bulk Order

Quality Level

form

liquid

mp

-25 °C

transition temp

flash point 105 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

assay n-C12 isomer (GC, area%): 60.0 - 70.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.873 - 0.875
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service