Skip to Content
Merck
All Photos(2)

Key Documents

8.00360

Sigma-Aldrich

2-(2-Aminoethylamino)ethanol

for synthesis

Synonym(s):

2-(2-Aminoethylamino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12N2O
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-867-5
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

345 °C

potency

3000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

3.3-10.1 % (v/v)

pH

11.8 (20 °C, 111 g/L in H2O)

bp

237-243 °C/1013 hPa

mp

-28 °C

transition temp

flash point 144 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H12N2O/c5-1-2-6-3-4-7/h6-7H,1-5H2

InChI key

LHIJANUOQQMGNT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.029 - 1.031
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(C)

144 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service