Skip to Content
Merck
All Photos(1)

Key Documents

8.00147

Sigma-Aldrich

Acetylenedicarboxylic acid

for synthesis

Synonym(s):

Acetylenedicarboxylic acid, Butynedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2O4
CAS Number:
Molecular Weight:
114.06
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-536-0
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

mp

180-187 °C

transition temp

flash point 131 °C

bulk density

550 kg/m3

InChI

1S/C4H2O4/c5-3(6)1-2-4(7)8/h(H,5,6)(H,7,8)

InChI key

YTIVTFGABIZHHX-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(C)

131 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service