Skip to Content
Merck
All Photos(1)

Key Documents

79109BC

Millipore

GDS PPMX Tip Disposal Bin

BioControl Systems, for use with GDS PickPen® PIPETMAX System

Synonym(s):

Pipette tip discard container, Pipette tip disposal bin, Pipette tip waste bin, Pipetting waste receptacle, Tip discard bin, Tip disposal container, Tip disposal unit, Tip waste container, Used tip collection bin, Used tip disposal bin

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$79.30

$79.30


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
1 EACH
$79.30

About This Item

UNSPSC Code:
41121521

$79.30


Estimated to ship on01 June 2025


Request more information

packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen® PIPETMAX System

storage temp.

room temp

Related Categories

General description

The GDS PPMX Tip Disposal Bin is a specialized bin designed for use with the GDS PickPen® PIPETMAX® (PPMX) System. It is used for the safe disposal of tips after they have been used during liquid handling. The disposal bin helps maintain a clean and organized workspace by providing a designated space to discard used tips, reducing the risk of cross-contamination.

Application

The GDS PPMX Tip Disposal Bin, an accessory used with the GDS PickPen® PIPETMAX® System, is primarily designed for the safe disposal of tips during liquid handling and pipetting processes.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service