Skip to Content
Merck
All Photos(1)

Key Documents

73184BC

Millipore

GDS PickPen II 8M Stand

BioControl Systems, for use with GDS PickPen®

Synonym(s):

GDS PickPen II 8M holder, GDS PickPen support, PickPen holder, Stand for GDS PickPen II 8M

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$627.00

$627.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
1 EACH
$627.00

About This Item

UNSPSC Code:
41115866

$627.00


Estimated to ship on29 May 2025


Request more information

packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen®

storage temp.

room temp

General description

GDS PickPen® II 8M Stand is a metal stand suitable to hold the manual GDS PickPen®, which is designed to transfer magnetic particles during Immuno Magnetic Separation (IMS) when working with Assurance GDS® pathogen detection kits. The stand is designed to hold up to three devices.

Application

GDS PickPen® II 8M Stand is a GDS PickPen® support accessory that is designed to hold the PickPen® during pathogen detection.

Legal Information

Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service