72622-M
NovaCHOice™ Transfection Kit
Optimized transfection reagent specifically developed for mammalian protein production in chinese hamster ovary (CHO) cells.
Synonym(s):
NovaCHOice Kit, Transfection Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
technique(s)
transfection: suitable
storage temp.
−20°C
Related Categories
General description
The NovaCHOice Transfection Kit consists of a NovaCHOice Transfection Reagent and NovaCHOice Booster Reagent specifically developed for mammalian protein production in Chinese Hamster Ovary (CHO) derived cells. The NovaCHOice Booster Reagent is optional and enhances expression in certain media formulations. Both reagents are comprised of animal-origin free components that are compatible with many chemically defined media formulations. The NovaCHOice Transfection Kit eliminates the need for a culture medium change post-transfection and is suitable for both transient and stable transfection.
Panel Components
• 1 x 1 ml, or 1 x 10 ml NovaCHOice Transfection Reagent
• 1 x 1 ml, or 1 x 10 ml NovaCHOice Booster Reagent
Storage: Store all components in a constant-temperature freezer at –20 °C. Before each use, warm to room temperature and vortex gently.
Panel Components
• 1 x 1 ml, or 1 x 10 ml NovaCHOice Transfection Reagent
• 1 x 1 ml, or 1 x 10 ml NovaCHOice Booster Reagent
Storage: Store all components in a constant-temperature freezer at –20 °C. Before each use, warm to room temperature and vortex gently.
Warning
Toxicity: Flammable (J)
Legal Information
NOVACHOICE is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service