Skip to Content
Merck
All Photos(1)

Key Documents

71340-M

Sigma-Aldrich

pCDFDuet-1 DNA - Novagen

Synonym(s):

Duet-1 DNA, Novagen DNA Vector, pCDFDuet-1 DNA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$755.00

$755.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 μG
$755.00

About This Item

UNSPSC Code:
41106602
NACRES:
NA.51

$755.00


Estimated to ship on29 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

Promoter

Promoter name: T7

selection

spectinomycin

storage temp.

−70°C

General description

pCDFDuet-1 is designed for the coexpression of two target genes. The vector encodes two multiple cloning sites (MCS) each of which is preceded by a T7 promoter, lac operator, and ribosome binding site (rbs). The vector also carries the pCloDF13 replicon, lacI gene and streptomycin/spectinomycin resistance marker. This vector can be transformed into the same cell with pETDuet-1, pACYCDuet-1, and pRSFDuet-1 or pCOLADuet-1 for the coexpression of up to eight target genes. ORFs inserted into MCS-1 can be sequenced using the ACYCDuetUP1 Primer and DuetDOWN1 Primer. ORFs inserted into MCS-2 can be sequenced using the DuetUP2 Primer and T7 Terminator Primer.

Application

pCDFDuet-1 DNA has been used as a vector for the expression of recombinant proteins in E-coli cells.[1][2][3]

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service