Skip to Content
Merck
All Photos(1)

Key Documents

71290-M

Millipore

5X AntigenPlus Buffer (pH 10)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$145.00

$145.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 ML
$145.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.77

$145.00


Estimated to ship on31 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

2-8°C

General description

Pretreatment of fixed tissues with an antigen retrieval buffer can dramatically improve antibody staining. For tissues and antibodies for which antigen retrieval is necessary, Novagen is pleased to introduce the AntigenPlus Buffer Set designed for antigen retrieval applications in fixed, paraffin embedded tissues. The Buffer Set consists of three different 5X AntigenPlus Buffers (pH 6, pH 7.4 and pH 10) that can be tested with the tissues of interest.

Warning

Toxicity: Irritant (B)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service