70920
Rosetta(DE3)pLacI Competent Cells - Novagen
Escherichia coli, rod shaped
About This Item
Recommended Products
Product Name
Rosetta(DE3)pLacI Competent Cells - Novagen, Rosetta strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. Contains the pLacI plasmid producing extra Lac repressor.
biological source
Escherichia coli
Quality Level
manufacturer/tradename
Novagen®
storage condition
OK to freeze
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
shipped in
dry ice
storage temp.
−70°C
1 of 4
This Item | 70954 | 70953 | 71058 |
---|---|---|---|
microbiological culture: suitable | microbiological culture: suitable | microbiological culture: suitable | microbiological culture: suitable |
Escherichia coli | Escherichia coli | Escherichia coli | Escherichia coli |
rod shaped | rod shaped | rod shaped | rod shaped |
−70°C | −70°C | −70°C | −70°C |
dry ice | wet ice | dry ice | dry ice |
General description
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Note that these strains are not recommended for use with pET, pACYCDuet, pCDF, pRSF, pCOLADuet or pETcoco vectors.
.Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) on a compatible chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters. In Rosetta(DE3)pLacI, the rare tRNA genes are present on the same plasmid that carries the lac repressor gene.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Components
•2 × 0.2 ml5 × 0.2 mlRosetta(DE3)pLacI Competent Cells
•2 × 2 ml4 × 2 mlSOC Medium
•2 ng2 ngTest Plasmid
Warning
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service