Skip to Content
Merck
All Photos(1)

Key Documents

70840-M

Sigma-Aldrich

pTriEx-1.1 DNA - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$860.00

$860.00


Estimated to ship on31 May 2025



Select a Size

Change View
20 μG
$860.00

About This Item

UNSPSC Code:
41106611
NACRES:
NA.51

$860.00


Estimated to ship on31 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

−20°C

General description

The pTriEx<TMSYMBOL></TMSYMBOL>-1.1 vector is uniquely designed to allow rapid characterization of target genes in multiple expression systems. With this vector a single recombinant plasmid can be used to test expression in E. coli, insect and vertebrate cells. Transient vertebrate expression is mediated by a hybrid promoter composed of the CMV immediate early enhancer fused to the chicken β-actin promoter. For expression in insect cells, pTriEx-1.1 contains flanking baculovirus sequences to permit the generation of recombinant baculoviruses using the BacVector<TMSYMBOL></TMSYMBOL> System. In baculovirus-infected insect cells, expression is driven by the very late p10 promoter. Expression in E. coli is regulated by the tightly controlled T7lac promoter. Expression can be induced in hosts such as NovaBlue by infecting with λCE6, a phage that constitutively expresses T7 RNA polymerase from the λpL and λ>pI promoters. Alternatively, pTriEx recombinant plasmids can be transferred into a (DE3)pLacI host that allows IPTG based induction.


This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].

Warning

Toxicity: Standard Handling (A)

Legal Information

This product is not available in Japan.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service