Skip to Content
Merck
All Photos(1)

Documents

70781

Sigma-Aldrich

pET Expression System 30 - Novagen

pET vectors plus host strains with induction control

Synonym(s):

pET-30a-c(+) Vectors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

conjugate

S-Tag

Quality Level

tag

His tagged

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

pET Expression Systems and pET
pET Expression Systems and pET Expression Systems plus Competent Cells provide core reagents needed for target gene cloning and expression.

Components: pET Expression Systems Components for pET Expression Systems are similar for all systems unless otherwise stated with the specific pET Expression System description. pET Expression Systems include:
  • 10 µg pET vector DNA (for each indicated plasmid)
  • 0.2 ml BL21 Glycerol Stock
  • ml BL21(DE3) Glycerol Stock
  • 0.2 ml BL21(DE3)pLysS Glycerol Stock
  • 0.2 ml Induction Control Glycerol Stock
Components: pET Expression Systems plus Competent Cells
pET Expression Systems plus Competent Cells contain all of the components of the specific pET Expression System, as well as the following additional components, unless otherwise stated with the specific pET Expression System description:
  • 0.2 ml NovaBlue Competent Cells
  • 0.2 ml BL21(DE3) Competent Cells
  • 0.2 ml BL21(DE3)pLysS Competent Cells
  • 2 × 0.2 ml SOC Medium
  • 10 µl Test Plasmid
These components are sufficient for up to 10 transformations in each host.

Purification and Detection Reagents
Purification and detection reagents are available separately. For complete product descriptions and ordering information, refer to the Protein Purification and Antibodies, Conjugates & Detection Tools chapters.

pET Expression System 3
The pET Expression System 3 contains 10 µg each of the four versions of pET-30 (pET-30a–c(+)). The pET-30a–c(+) vectors carry an N-terminal His•Tag®/thrombin/S•Tag/enterokinase configuration plus an optional C-terminal His•Tag sequence. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map, TB095VM.

Components

  • 10 µg - eachpET-30a-c(+) vector DNA
  • 0.2 ml- Host bacterial strains BL21, BL21(DE3), and BL21(DE3)pLysS, glycerol stocks
  • 0.2 ml- Induction control clone, glycerol stock

Warning

Toxicity: Standard Handling

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service