69752
pET-24d(+) DNA - Novagen
Synonym(s):
pET-24a-d(+) vectors, pET-24d(+) DNA vectors
Select a Size
About This Item
Recommended Products
manufacturer/tradename
Novagen®
Quality Level
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
dry ice
wet ice
storage temp.
−70°C
Related Categories
General description
Application
Warning
Other Notes
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.
Legal Information
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service