Skip to Content
Merck
All Photos(1)

Key Documents

69048

Sigma-Aldrich

T7 Tag Antibody HRP Conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$663.00

$663.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$663.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

$663.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service