Skip to Content
Merck
All Photos(1)

Key Documents

676491

Sigma-Aldrich

VEGF Inducer, GS4012

A cell-permeable pyridinyl-thioether compound that acts as a potent inducer of VEGF and VEGF-mediated vessel formation.

Synonym(s):

VEGF Inducer, GS4012, 4-(2-(4-Methoxyphenylsulfanyl)ethyl)pyridine, HCl, GS4012

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$293.00

$293.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$293.00

About This Item

Empirical Formula (Hill Notation):
C14H15NOS · xHCl
CAS Number:
Molecular Weight:
245.34 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$293.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 200 mg/mL
methanol: 50 mg/mL
water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

S(CCc2ccncc2)c1ccc(cc1)OC.Cl

InChI

1S/C14H15NOS.ClH/c1-16-13-2-4-14(5-3-13)17-11-8-12-6-9-15-10-7-12;/h2-7,9-10H,8,11H2,1H3;1H

InChI key

CYZALCLQLHDAEC-UHFFFAOYSA-N

General description

A cell-permeable and potent inducer of VEGF and VEGF-mediated blood vessel formation. GS4012-induced upregulation of VEGF correlates with its ability to suppress the gridlock/coarctation phenotype (7.5 µg/ml) in Zebrafish and to stimulate tubule network formation (5 µg/ml) in HUVECs. An essential molecule required during angioblasts cell fate determination.
A cell-permeable pyridinyl-thioether compound that acts as a potent inducer of VEGF and VEGF-mediated vessel formation. GS4012-induced upregulation of VEGF correlates well with its ability to suppress the gridlock/coarctation phenotype (7.5 µg/ml) in Zebrafish and to stimulate tubule network formation (5 µg/ml) in HUVECs.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Potent inducer of VEGF and VEGF-mediated vessel formation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Peterson, R.T., et al. 2004. Nat. Biotechnol.22, 595.
Depre, C., et al. 2003. Circulation108, 2723.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service