Skip to Content
Merck
All Photos(1)

Key Documents

662041

Sigma-Aldrich

U-73343

A cell-permeable analog of U-73122 that acts as a very weak inhibitor of phospholipase C. Suitable as a negative control.

Synonym(s):

U-73343, 1-[6-((17β-3-Methoxyestra-1,3,5(10)-trien-17-yl)amino)hexyl]-2,5-pyrrolidinedione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$232.00

$232.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 MG
$232.00

About This Item

Empirical Formula (Hill Notation):
C29H42N2O3
CAS Number:
Molecular Weight:
466.66
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$232.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

ethanol: 1 mg/mL
DMSO: 2 mg/mL
chloroform: 200 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

N5(C(=O)CCC5=O)CCCCCCN[C@@H]1[C@@]2([C@H]([C@H]3[C@H](CC2)c4c(cc(cc4)OC)CC3)CC1)C

InChI

1S/C29H42N2O3/c1-29-16-15-23-22-10-8-21(34-2)19-20(22)7-9-24(23)25(29)11-12-26(29)30-17-5-3-4-6-18-31-27(32)13-14-28(31)33/h8,10,19,23-26,30H,3-7,9,11-18H2,1-2H3/t23-,24-,25+,26+,29+/m1/s1

InChI key

CJHWFIUASFBCKN-ZRJUGLEFSA-N

General description

A cell-permeable analog of U-73122 (Cat. No. 662035) that acts as a very weak inhibitor of phospholipase C. Suitable as a negative control.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Negative control of U-73122 in the inhibition of phospholipase C
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

For storage in chloroform, aliquot into single use volumes, evaporate to dryness under a stream of nitrogen and freeze (-20°C). DMSO stocks solutions are stable for up to 6 months at -20°C.

Other Notes

Tatrai, A., et al. 1994. Biochim. Biophys. Acta 1224, 595.
Bleasdale, J.E., et al. 1990. J. Pharmacol. Exp. Ther.255, 756.
Smith, R.J., et al. 1990. J. Pharmacol. Exp. Ther.253, 688.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Norman et al.
FEBS letters, 484(3), 179-183 (2000-11-18)
Aggregation by immune complexes of receptors specific for the Fc region of IgG results in their internalisation and disposal by trafficking to lysosomes. We show here that internalisation of FcgammaRI by IFN-gamma treated U937 cells following receptor aggregation by cross-linking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service