Skip to Content
Merck
All Photos(1)

Key Documents

65092-M

Sigma-Aldrich

Gram Stain Set

Synonym(s):

bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

application(s)

diagnostic assay manufacturing
hematology
histology

Quality Level

storage temp.

15-25°C

Application

HARLECO® Complete bacterial staining kit (4 x 500mL) containing crystal violet, iodine, decolorizer and safranin stain.
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For use in Gram staining of bacteria.

Components

Complete kit containing crystal violet, iodine, decolorizer and safranin stain.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Central nervous system, Thyroid

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service