Skip to Content
Merck
All Photos(1)

Key Documents

648465

Millipore

Triton X-100 Detergent, Hydrogenated

Nonionic detergent used for the solubilization of membrane-bound proteins.

Synonym(s):

Octylphenoxypolyethoxyethanol, Hydrogenated, TRX-100

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$525.00

$525.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$525.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

$525.00


Estimated to ship on20 May 2025


Request a Bulk Order

description

Merck USA index - 14, 6761

Quality Level

form

viscous liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless

CMC

250 μM

solubility

water: 10%

shipped in

ambient

storage temp.

15-25°C

SMILES string

O(C1CCC(CC1)C(CC(C)(C)C)(C)C)CCOCCOCCOCCOCCOCCOCCO

InChI

1S/C28H56O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h25-26,29H,6-24H2,1-5H3

InChI key

QQJNBKDKLMCALZ-UHFFFAOYSA-N

General description

Nonionic detergent used for the solubilization of membrane-bound proteins. Lacks the UV absorbance and fluorescence of TRITON X-100 (Cat. No. 648462), which causes interference in spectophotometric monitoring and assays. Has a critical micelle concentration (CMC) of 250 µM.

Application


  • The utilization of Triton X-100 for enhanced two-dimensional liquid-phase proteomics.: This study explores the role of Triton X-100 in enhancing protein separation techniques in biochemistry, particularly emphasizing its application in two-dimensional liquid-phase proteomics to improve resolution and reproducibility (Kim et al., 2011).

  • Effects of environment on the photophysical characteristics of mesotetrakis methylpyridiniumyl porphyrin (TMPyP).: Research focuses on the influence of different solvent environments, including those containing Triton X-100, on the photophysical properties of porphyrins, highlighting its utility in enhancing solubility and stability in biochemical assays (Gonccedil;alves et al., 2011).

  • Processing of tissue culture cells.: The work describes methods for processing tissue culture cells where Triton X-100 is used to permeabilize cells for various staining procedures, crucial for molecular biology and cellular biochemistry studies (Bratthauer, 2010).

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following dilution in H₂O, store at room temperature. Diluted stock solutions are stable for up to 3 months at room temperature.

Other Notes

Fernandez, J., et al. 1992. Anal. Biochem.201, 255.
Tiller, G.E., et al. 1984. Anal. Biochem.141, 262.
Ashani, Y. and Cantravas, G.N. 1980. Anal. Biochem.109, 55.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service