Skip to Content
Merck
All Photos(1)

Documents

648462

Millipore

Triton X-100

Non-ionic detergent and emulsifier

Synonym(s):

Triton X-100, Polyethylene Glycol- p-isooctylphenyl Ether, Octylphenoxypolyethoxyethanol, Polyethylene Glycol-p-isooctylphenyl Ether, Octylphenoxypolyethoxyethanol, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

product name

Triton X-100 Detergent, Non-ionic detergent and emulsifier. Aggregation number: 100-155.

Quality Level

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

100-155

color

colorless to pale yellow

CMC

0.2-0.9 mM (20-25°C)

transition temp

cloud point 65 °C
pour point ~7 °C

solubility

water: soluble

HLB

13.5

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HLB 13.5. Cloud point 65°C.
Non-ionic detergent and emulsifier (aggregation number:100-55; cloud point: 65°C; CMC: 200-900 µM; HLB number: 13.5).
Non-ionic detergent and emulsifier. Aggregation number: 100-155.

Application


  • Barbeya oleoides Leaves Extracts: In Vitro Carbohydrate Digestive Enzymes Inhibition and Phytochemical Characterization.: Though indirectly related, this study utilizes methodologies where Triton X-100′s properties as a detergent can be crucial for the extraction and stabilization of bioactive compounds from plant sources, important for medicinal chemistry applications (Khojah et al., 2021).

  • Surface Plasmon Resonance Monitoring of Mono-Rhamnolipid Interaction with Phospholipid-Based Liposomes.: Explores the interaction of biomolecules with lipid membranes, where Triton X-100 plays a key role in forming and stabilizing these liposomes, essential for biochemists studying membrane dynamics and drug delivery systems (Belkilani et al., 2021).

  • Sampling the cultivation parameter space for the bacterial production of TLR1 intracellular domain reveals the multiple optima.: Discusses the use of Triton X-100 in protein purification processes, underlining its utility in optimizing conditions for the production of biologically active proteins, a common technique in biochemical research labs (Goncharuk et al., 2021).

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service