Skip to Content
Merck
All Photos(2)

Key Documents

648310-M

Millipore

Tris Base, Molecular Biology Grade

≥99% (perchloric acid titration), solid, pH 7.0-9.0, solid, Molecular Biology Grade

Synonym(s):

Tris(hydroxymethyl)aminomethane, tris(Hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$254.00
2.5 KG
$762.00

$254.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 G
$254.00
2.5 KG
$762.00

About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.25

$254.00


In StockDetails


Request a Bulk Order

Product Name

Tris Base, Molecular Biology Grade, TRIS base is useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

grade

Molecular Biology

Quality Level

Assay

≥99% (perchloric acid titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

water: 500 mg/mL

cation traces

heavy metals: ≤0.0005%

absorption

≤0.02 at 280 nm
≤0.03 at 260 nm

foreign activity

RNases, DNases, proteases, none detected

storage temp.

10-30°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.
Widely used as a biological buffer (absorbance: ≤0.03 at 260 nm; ≤0.02 at 280 nm). Useful in the pH range of 7.0-9.0. Has a pKa of 8.1 at 25°C.

Application


  • A Fast and Automated Melanin-bleaching Method for Histopathologic Evaluation of Pigmented Melanoma Tissues.: This study presents an automated melanin-bleaching technique using a Tris-based buffer to enhance histopathologic evaluation of pigmented melanoma tissues, aiding in more accurate diagnoses (Ugolini et al., 2022).

  • Improvement of plant protein solubilization and 2-DE gel resolution through optimization of the concentration of Tris in the solubilization buffer.: Highlights the optimization of Tris concentration in solubilization buffers to enhance protein solubilization and gel electrophoresis resolution, crucial for biochemical studies (Cho et al., 2010).

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service