Skip to Content
Merck
All Photos(2)

Documents

6442

Sigma-Aldrich

Water

Deionized Distilled, ASTM Type II, HARLECO® Purified water for laboratory use

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
41116121

product name

Water, Deionized Distilled, ASTM Type II, HARLECO® Purified water for laboratory use

vapor pressure

23 hPa ( 20 °C)

form

liquid

shelf life

18 mo. (from manufacture date)

color

APHA: ≤10

refractive index

n20/D 1.34 (lit.)
n20/D 1.34

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)
1.000 g/mL at 3.98 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HARLECO® Purified water for laboratory use

Analysis Note

Expiration date18 months from mfg dateChloride (Cl)5ppb maxColor (APHA)10APHA maxConductivity at 25°C1.0µS/cm maxFormClear liquidHeavy metals (as Pb)0.01ppm maxIon chromatography, Chloride0.4ppm maxIon chromatography, Nitrate 0.4ppm maxIon chromatography, Phosphate 1.0ppm maxIon chromatography, Sulfate 1.0ppm maxIron (Fe)0.3ppm maxMicrobial limits10CFU/mL maxResistivity at 25°C1.0MOhm·cm minSilica (SiO2)3ppb maxSodium (Na)5ppb maxSubstances reducing permanganateTo pass testTotal organic carbon50µg/L maxTotal solids20ppm max

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service