Recommended Products
species reactivity
all
technique(s)
western blot: suitable
detection method
chemiluminescent
General description
The Visualizer Western Blot Detection Kit is designed for the sensitive detection of proteins in chemiluminescent Western blots.
Application
For use on immunoblots.
Packaging
Kit capacity: 20 mini-gel-sized blots for 1000 cm2 of membrane
Components
Goat Anti-Mouse IgG, HRP conjugate (Cat.# 12-349) Visualizer Detection Reagent A Visualizer Detection Reagent B
Quality
The reagents in this kit have been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.
Storage and Stability
Stable for 18 months at 4°C from date of shipment. Note: Upon receipt, resuspend, aliquot and store Catalog # 12-349 at -20°C for optimal performance. Store Visualizer Detection Reagents at 4°C.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service