Skip to Content
Merck
All Photos(1)

Key Documents

6350-OP

Millipore

MOPS

OmniPur® Grade, 10X Concentrate solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$271.00

$271.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$271.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

$271.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

color

clear to haze-free colorless

shipped in

ambient

General description

Ideal for RNA electrophoresis in agarose. Buffering range 6.5-7.9. Each liter of concentrate makes 10L of 1X MOPS. Prepared with MOPS and DEPC for RNA applications. Molecular biology grade.

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the concentration of the solution, in Molar or mM ?

    1 answer
    1. The concentration of this product is listed in the corresponding Safety Data Sheet as follows:
      "OmniPur® 10X MOPS (200 mM MOPS, 50 mM Sodium Acetate, 10 mM EDTA, 0.1% DEPC"

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service