Skip to Content
Merck
All Photos(1)

Documents

627624

Sigma-Aldrich

D-Val-Phe-Lys Chloromethyl Ketone, Dihydrochloride

Selective irreversible inhibitor of plasmin with high selectivity for plasmin (IC₅₀ = 100 pM for human plasmin) over urokinase.

Synonym(s):

Plasmin Inhibitor, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Selective irreversible inhibitor of plasmin with high selectivity for plasmin (IC50 = 100 pM for human plasmin) over urokinase.
Selective irreversible inhibitor of plasmin with high selectivity for plasmin over urokinase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Human plasmin
Product does not compete with ATP.
Reversible: no
Target IC50: 100 pM for human plasmin

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-D-Val-Phe-Lys-CMK

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.







From Catalog:

Desc. Field- added "(IC₅₀ = 100 pM for human plasmin) "

Other Notes

Woessner, J.F., Jr., et al. 1989. Steroids54, 491.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasam Madarati et al.
Scientific reports, 14(1), 9073-9073 (2024-04-21)
ADAMTS13, a disintegrin and metalloprotease with a thrombospondin type 1 motif, member 13, regulates the length of Von Willebrand factor (VWF) multimers and their platelet-binding activity. ADAMTS13 is constitutively secreted as an active protease and is not inhibited by circulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service